2,6-Dichloro-3-fluoroisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

62029-1G 62029-25G 62029-500MG 62029-5G
76905-490EA 215.37 USD
76905-490 76905-492 76905-494 76905-496
2,6-Dichloro-3-fluoroisonicotinic acid ≥97%
2,6-Dichloro-3-fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24490307
CAS Number: 149468-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR