2-Chloro-N-methylisonicotinamide ≥95%

Supplier: AOBChem USA

Warning

60957-1G 60957-25G 60957-5G
76905-434EA 73.21 USD
76905-434 76905-436 76905-438
2-Chloro-N-methylisonicotinamide ≥95%
2-Chloro-N-methylisonicotinamide

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09862946
CAS Number: 131418-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR