2-Iodo-4-methoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60221-1G 60221-25G 60221-500MG 60221-5G
76905-384EA 296 USD
76905-384 76905-386 76905-388 76905-390
2-Iodo-4-methoxy-1-methylbenzene ≥97%
2-Iodo-4-methoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₉IO
MW: 248.061 g/mol
Storage Temperature: Refrigerator
CAS Number: 260558-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR