2-Ethoxy-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60123-1G 60123-25G 60123-500MG 60123-5G
76905-372EA 129.43 USD
76905-372 76905-374 76905-376 76905-378
2-Ethoxy-3-methoxybenzoic acid ≥97%
2-Ethoxy-3-methoxybenzoic acid
Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD06203281
CAS Number: 773869-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR