2,4-Difluoro-5-methylphenol ≥95%

Supplier: AOBChem USA

Warning

59212-1G 59212-25G 59212-500MG 59212-5G
76905-340EA 405.27 USD
76905-340 76905-342 76905-344 76905-346
2,4-Difluoro-5-methylphenol ≥95%
2,4-Difluoro-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MW: 144.119 g/mol
Storage Temperature: Ambient
CAS Number: 1378584-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR