2-Chloro-3,6-difluorophenetole 97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-3-ethoxy-1,4-difluorobenzene

58959-1G 58959-25G 58959-5G 58959-500MG
76905-326EA 84.88 USD
76905-326 76905-328 76905-330 77651-782
2-Chloro-3,6-difluorophenetole 97% research grade
2-Chloro-3,6-difluorophenetole
Formula: C₈H₇ClF₂O
MW: 192.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28750542
CAS Number: 1807166-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR