2-Bromo-6-chloro-4-ethylaniline ≥97%

Supplier: AOBChem USA

Warning

58398-1G 58398-25G 58398-500MG 58398-5G
76905-300EA 296 USD
76905-300 76905-302 76905-304 76905-306
2-Bromo-6-chloro-4-ethylaniline ≥97%
2-Bromo-6-chloro-4-ethylaniline

Precaución: for Research Use Only

Formula: C₈H₉BrClN
MW: 234.521 g/mol
Storage Temperature: Refrigerator
CAS Number: 2090692-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR