2-Bromo-1-chloro-5-methoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

56985-1G 56985-25G 56985-500MG 56985-5G
76905-262EA 233.4 USD
76905-262 76905-264 76905-266 76905-268
2-Bromo-1-chloro-5-methoxy-3-methylbenzene ≥95%
2-Bromo-1-chloro-5-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
CAS Number: 2091663-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR