2,6-Difluoro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56935-1G 56935-25G 56935-500MG 56935-5G
76905-250EA 129.43 USD
76905-250 76905-252 76905-254 76905-256
2,6-Difluoro-4-iodobenzaldehyde ≥97%
2,6-Difluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD11845966
CAS Number: 1160573-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR