2-Formylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

56844-1G 56844-25G 56844-500MG 56844-5G
76905-234EA 233.4 USD
76905-234 76905-236 76905-238 76905-240
2-Formylnicotinic acid ≥97%
2-Formylnicotinic acid
Formula: C₇H₅NO₃
MW: 151.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20642387
CAS Number: 23590-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR