2-Iodo-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56605-1G 56605-25G 56605-500MG 56605-5G
76905-218EA 135.8 USD
76905-218 76905-220 76905-222 76905-224
2-Iodo-5-methylbenzaldehyde ≥97%
2-Iodo-5-methylbenzaldehyde
Formula: C₈H₇IO
Storage Temperature: Refrigerator
MDL Number: MFCD18397772
CAS Number: 1106813-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR