2-Fluoro-3-iodophenol ≥95%

Supplier: AOBChem USA

Warning

56375-1G 56375-25G 56375-500MG 56375-5G
76905-204EA 165.51 USD
76905-204 76905-206 76905-208 76905-210
2-Fluoro-3-iodophenol ≥95%
2-Fluoro-3-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
Storage Temperature: Ambient
MDL Number: MFCD12910496
CAS Number: 897957-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR