2-Fluoro-3-iodo-6-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

55389-1G 55389-25G 55389-500MG 55389-5G
76905-154EA 379.81 USD
76905-154 76905-156 76905-158 76905-160
2-Fluoro-3-iodo-6-methoxypyridine ≥97%
2-Fluoro-3-iodo-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅FINO
MW: 253.013 g/mol
Storage Temperature: Refrigerator
CAS Number: 1227177-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR