2-Bromo-6-chloronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

55185-1G 55185-25G 55185-500MG 55185-5G
76905-138EA 375.56 USD
76905-138 76905-140 76905-142 76905-144
2-Bromo-6-chloronicotinic acid ≥97%
2-Bromo-6-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185791
CAS Number: 1060815-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR