2,5-Dichloro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54783-1G 54783-25G 54783-500MG 54783-5G
76905-080EA 346.92 USD
76905-080 76905-082 76905-084 76905-086
2,5-Dichloro-4-methoxybenzaldehyde ≥97%
2,5-Dichloro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13192728
CAS Number: 381229-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR