2-Iodo-6-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

54729-1G 54729-500MG 54729-5G
76905-066EA 643.97 USD
76905-066 76905-068 76905-070
2-Iodo-6-methoxyphenol ≥97% research grade
2-Iodo-6-methoxyphenol
Formula: C₇H₇IO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11054827
CAS Number: 111726-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR