2-Bromo-5-nitrobenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
54366-1G
54366-25G
54366-500MG
54366-5G
76905-050EA
147.47
USD
76905-050
76905-052
76905-054
76905-056
2-Bromo-5-nitrobenzamide ≥97%
2-Bromo-5-nitrobenzamide
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂O₃ MW: 245.03 g/mol Storage Temperature: Refrigerator |
CAS Number:
41052-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...