2,5-Dichloro-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

54202-1G 54202-25G 54202-500MG 54202-5G
76905-034EA 381.93 USD
76905-034 76905-036 76905-038 76905-040
2,5-Dichloro-3-methylaniline ≥97%
2,5-Dichloro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD20700007
CAS Number: 1379291-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR