2-Methoxy-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53868-25G 53868-5G
76905-000EA 456.19 USD
76905-000 76905-002
2-Methoxy-4-(trifluoromethoxy)benzaldehyde ≥97%
2-Methoxy-4-(trifluoromethoxy)benzaldehyde
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115965
CAS Number: 886500-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR