2-Chloro-4-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

53576-1G 53576-25G 53576-500MG 53576-5G
76904-974EA 292.81 USD
76904-974 76904-976 76904-978 76904-980
2-Chloro-4-isopropylphenol ≥97%
2-Chloro-4-isopropylphenol
Formula: C₉H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD16997172
CAS Number: 51202-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR