2-Bromo-4-fluoro-5-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
53545-25G
53545-5G
76904-962EA
418
USD
76904-962
76904-964
2-Bromo-4-fluoro-5-methylbenzoic acid ≥97%
2-Bromo-4-fluoro-5-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09263434 CAS Number: 1003709-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...