2-Bromo-5-iodobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

53396-25G 53396-5G
76904-938EA 453.01 USD
76904-938 76904-940
2-Bromo-5-iodobenzonitrile ≥97%
2-Bromo-5-iodobenzonitrile
Formula: C₇H₃BrIN
Storage Temperature: Ambient
MDL Number: MFCD09025663
CAS Number: 1252046-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR