2-Bromo-1-fluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53231-1G 53231-25G 53231-500MG 53231-5G
76904-916EA 292.81 USD
76904-916 76904-918 76904-920 76904-922
2-Bromo-1-fluoro-3-isopropoxybenzene ≥97%
2-Bromo-1-fluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD28356392
CAS Number: 1369905-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR