2-Iodo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

53058-1G 53058-25G 53058-500MG 53058-5G
76904-880EA 242.95 USD
76904-880 76904-882 76904-884 76904-886
2-Iodo-5-methylphenol ≥97%
2-Iodo-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₇IO
MW: 234.034 g/mol
Storage Temperature: Refrigerator
CAS Number: 35928-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR