2-Chloro-4-methyl-6-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

52987-1G 52987-25G 52987-500MG 52987-5G
76904-866EA 260.99 USD
76904-866 76904-868 76904-870 76904-872
2-Chloro-4-methyl-6-(methylthio)pyridine ≥97%
2-Chloro-4-methyl-6-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNS
MW: 173.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404103
CAS Number: 2635937-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR