2-Chloro-6-ethylpyridine ≥97%

Supplier: AOBChem USA

Warning

52749-1G 52749-500MG 52749-5G
76904-838EA 638.67 USD
76904-838 76904-840 76904-842
2-Chloro-6-ethylpyridine ≥97%
2-Chloro-6-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
Storage Temperature: Ambient
MDL Number: MFCD11043821
CAS Number: 109201-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR