2-Bromo-4-ethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

52530-1G 52530-25G 52530-500MG 52530-5G
76904-822EA 260.99 USD
76904-822 76904-824 76904-826 76904-828
2-Bromo-4-ethylbenzoic acid ≥95%
2-Bromo-4-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18410373
CAS Number: 51605-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR