2-Bromo-4-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

52422-1G 52422-25G 52422-500MG 52422-5G
76904-796EA 379.81 USD
76904-796 76904-798 76904-800 76904-802
2-Bromo-4-fluoro-1,1'-biphenyl ≥97%
2-Bromo-4-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrF
Storage Temperature: Refrigerator
MDL Number: MFCD03411552
CAS Number: 53591-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR