2-Fluoro-6-hydroxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52298-1G 52298-25G 52298-500MG 52298-5G
76904-782EA 292.81 USD
76904-782 76904-784 76904-786 76904-788
2-Fluoro-6-hydroxy-4-methylbenzoic acid ≥97%
2-Fluoro-6-hydroxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33404037
CAS Number: 2386220-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR