2-Bromo-1-fluoro-3-(methoxymethoxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52287-1G 52287-25G 52287-500MG 52287-5G
76904-766EA 203.7 USD
76904-766 76904-768 76904-770 76904-772
2-Bromo-1-fluoro-3-(methoxymethoxy)-4-methylbenzene ≥97%
2-Bromo-1-fluoro-3-(methoxymethoxy)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33404033
CAS Number: 2624417-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR