2-Chloro-3',4'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52188-1G 52188-500MG 52188-5G
76904-710EA 589.87 USD
76904-710 76904-712 76904-714
2-Chloro-3',4'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2-Chloro-3',4'-difluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇ClF₂O
MW: 252.644 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32111808
CAS Number: 1962207-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR