2-Fluoro-3-iodo-4-methoxy-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

51522-1G 51522-25G 51522-5G
76904-672EA 117.76 USD
76904-672 76904-674 76904-676
2-Fluoro-3-iodo-4-methoxy-1-methylbenzene ≥95%
2-Fluoro-3-iodo-4-methoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD32902951
CAS Number: 2387271-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR