2-Chloro-4-fluoro-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

51331-1G 51331-500MG 51331-5G
76904-656EA 528.33 USD
76904-656 76904-658 76904-660
2-Chloro-4-fluoro-1,3-dimethylbenzene ≥97%
2-Chloro-4-fluoro-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClF
MW: 158.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751483
CAS Number: 26829-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR