2-Fluoro-3,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

50776-1G 50776-25G 50776-500MG 50776-5G
76904-616EA 262.05 USD
76904-616 76904-618 76904-620 76904-622
2-Fluoro-3,4-dimethoxybenzoic acid ≥97%
2-Fluoro-3,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₄
Storage Temperature: Refrigerator
MDL Number: MFCD19439835
CAS Number: 2967-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR