2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

50740-1G 50740-25G 50740-500MG 50740-5G
76904-602EA 342.68 USD
76904-602 76904-604 76904-606 76904-608
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzaldehyde ≥95%
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27975486
CAS Number: 1823327-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR