2-Fluoro-3-hydroxy-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49863-1G 49863-25G 49863-500MG 49863-5G
76904-544EA 379.81 USD
76904-544 76904-546 76904-548 76904-550
2-Fluoro-3-hydroxy-4-methoxybenzoic acid ≥97%
2-Fluoro-3-hydroxy-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₄
Storage Temperature: Ambient
MDL Number: MFCD11520919
CAS Number: 1260784-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR