2,5-Dibromo-3-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dibromo-1-methyl-3-nitrobenzene

49626-1G 49626-25G 49626-500MG 49626-5G
76904-514EA 183.54 USD
76904-514 76904-516 76904-518 76904-520
2,5-Dibromo-3-nitrotoluene ≥97%
2,5-Dibromo-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11846032
CAS Number: 408340-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR