2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49586-1G 49586-25G 49586-500MG 49586-5G
76904-498EA 409.51 USD
76904-498 76904-500 76904-502 76904-504
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₅H₂₁BO₃
MW: 260.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902841
CAS Number: 2828440-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR