2-Bromo-6-isobutoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

49522-25G 49522-5G
76904-488EA 595.17 USD
76904-488 76904-490
2-Bromo-6-isobutoxynaphthalene ≥97%
2-Bromo-6-isobutoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₄H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD18262544
CAS Number: 1363386-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR