2-Bromo-4-ethoxy-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

49447-1G 49447-25G 49447-500MG 49447-5G
76904-476EA 157.02 USD
76904-476 76904-478 76904-480 76904-482
2-Bromo-4-ethoxy-1-fluorobenzene ≥97%
2-Bromo-4-ethoxy-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25318992
CAS Number: 1555379-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR