2-Fluoro-3-formyl-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49351-1G 49351-25G 49351-500MG 49351-5G
76904-458EA 168.69 USD
76904-458 76904-460 76904-462 76904-464
2-Fluoro-3-formyl-5-methylbenzoic acid ≥97%
2-Fluoro-3-formyl-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃
MW: 182.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403933
CAS Number: 2167871-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR