2-Bromo-4-chloro-5-methylaniline ≥97%

Supplier: AOBChem USA

Warning

49196-1G 49196-25G 49196-500MG 49196-5G
76904-428EA 434.97 USD
76904-428 76904-430 76904-432 76904-434
2-Bromo-4-chloro-5-methylaniline ≥97%
2-Bromo-4-chloro-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.494 g/mol
Storage Temperature: Ambient
CAS Number: 939989-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR