2-Chloro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49105-25G 49105-5G
76904-418EA 346.92 USD
76904-418 76904-420
2-Chloro-5-hydroxybenzaldehyde ≥97%
2-Chloro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07783652
CAS Number: 7310-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR