2,6-Difluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-3-tolualdehyde, 2,6-Difluoro-m-tolualdehyde, 2,6-Difluoro-m-tolylaldehyde, 2,6-Difluoro-3-tolylaldehyde

49055-25G 49055-5G
76904-410EA 254.62 USD
76904-410 76904-412
2,6-Difluoro-3-methylbenzaldehyde ≥97%
2,6-Difluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631333
CAS Number: 261763-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR