2-Fluoro-5-iodo-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48891-1G 48891-25G 48891-500MG 48891-5G
76904-384EA 129.43 USD
76904-384 76904-386 76904-388 76904-390
2-Fluoro-5-iodo-4-methylbenzoic acid ≥97%
2-Fluoro-5-iodo-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107264
CAS Number: 1300741-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR