2-Bromo-4-fluoro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48848-1G 48848-25G 48848-500MG 48848-5G
76904-368EA 263.11 USD
76904-368 76904-370 76904-372 76904-374
2-Bromo-4-fluoro-5-hydroxybenzaldehyde ≥97%
2-Bromo-4-fluoro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
CAS Number: 1262989-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR