2-Chloro-4-iodopyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

48635-1G 48635-25G 48635-500MG 48635-5G
76904-342EA 189.91 USD
76904-342 76904-344 76904-346 76904-348
2-Chloro-4-iodopyridin-3-ol ≥97%
2-Chloro-4-iodopyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃ClINO
MW: 255.441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610456
CAS Number: 1207973-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR