2-Fluoro-3-hydroxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

48399-1G 48399-25G 48399-500MG 48399-5G
76904-296EA 292.81 USD
76904-296 76904-298 76904-300 76904-302
2-Fluoro-3-hydroxyisonicotinic acid ≥95%
2-Fluoro-3-hydroxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₃
MW: 157.099 g/mol
Storage Temperature: Ambient
CAS Number: 1227576-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR