2-Chloro-4,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48328-1G 48328-25G 48328-500MG 48328-5G
76904-248EA 124.13 USD
76904-248 76904-250 76904-252 76904-254
2-Chloro-4,5-dimethoxybenzaldehyde ≥97%
2-Chloro-4,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00461433
CAS Number: 18093-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR