2-Bromo-1-chloro-5-fluoro-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

48275-1G 48275-500MG 48275-5G
76904-236EA 589.87 USD
76904-236 76904-238 76904-240
2-Bromo-1-chloro-5-fluoro-3-(trifluoromethyl)benzene ≥95%
2-Bromo-1-chloro-5-fluoro-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂BrClF₄
MW: 277.441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD9033156
CAS Number: 914225-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR